The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000414.3 | NP_000405.1 | 7 - 8 | 565 | 118 |
NM_001199291.2 | NP_001186220.1 | 8 - 9 | 764 | 118 | |
NM_001199292.1 | NP_001186221.1 | 6 - 7 | 511 | 118 | |
NM_001292027.1 | NP_001278956.1 | 8 - 9 | 630 | 118 | |
GenBank mRNA | AK298075.1 | - | 8 - 9 | 692 | 118 |
AK299685.1 | - | 8 - 9 | 535 | 118 | |
AK301212.1 | - | 6 - 7 | 457 | 118 | |
AK308968.1 | - | 2 - 3 | 181 | 118 | |
AK312449.1 | - | 7 - 8 | 511 | 118 | |
BC003098.1 | - | 7 - 8 | 499 | 118 | |
KU178135.1 | - | 7 - 8 | 432 | 118 | |
X87176.1 | - | 7 - 8 | 480 | 118 |