The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052911.2 | NP_443143.2 | 11 - 12 | 3312 | 108 |
XM_011525798.1 | XP_011524100.1 | 11 - 12 | 3442 | 108 | |
XM_011525799.2 | XP_011524101.1 | 10 - 11 | 3485 | 108 | |
GenBank mRNA | AB067498.1 | - | 11 - 12 | 3248 | 108 |
AK090579.1 | - | 8 - 9 | 1070 | 108 | |
AL832041.1 | - | 12 - 13 | 3588 | 108 | |
AL834200.1 | - | 9 - 10 | 2144 | 108 | |
BC036943.1 | - | 5 - 6 | 681 | 108 | |
BC089426.1 | - | 8 - 9 | 2732 | 108 |