The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052911.2 | NP_443143.2 | 9 - 10 | 2967 | 147 |
XM_011525798.1 | XP_011524100.1 | 9 - 10 | 3097 | 147 | |
GenBank mRNA | AB067498.1 | - | 9 - 10 | 2903 | 147 |
AK090579.1 | - | 6 - 7 | 725 | 147 | |
BC089426.1 | - | 6 - 7 | 2387 | 147 |