The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006909.2 | NP_008840.1 | 24 - 25 | 3489 | 133 |
XM_017009682.1 | XP_016865171.1 | 24 - 25 | 3171 | 133 | |
GenBank mRNA | AF023130.1 | - | 24 - 25 | 3439 | 133 |
AF181250.1 | - | 24 - 25 | 3439 | 133 | |
BC126112.1 | - | 24 - 25 | 3489 | 133 | |
BC136296.1 | - | 24 - 25 | 3489 | 133 |