The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006909.2 | NP_008840.1 | 9 - 10 | 1441 | 61 |
XM_005248565.1 | XP_005248622.1 | 9 - 10 | 1458 | 61 | |
XM_017009682.1 | XP_016865171.1 | 9 - 10 | 1123 | 61 | |
XM_017009683.1 | XP_016865172.1 | 9 - 10 | 1458 | 61 | |
GenBank mRNA | AF023130.1 | - | 9 - 10 | 1391 | 61 |
AF181250.1 | - | 9 - 10 | 1391 | 61 | |
BC126112.1 | - | 9 - 10 | 1441 | 61 | |
BC136296.1 | - | 9 - 10 | 1441 | 61 | |
BC143870.1 | - | 9 - 10 | 1441 | 61 |