Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK4
Entrez Gene ID
6789
Gene Name
serine/threonine kinase 4
Gene Aliases
KRS2, MST1, YSK3
Chromosome Location
Chr.20: 44966474 - 45079977 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006282.3 NP_006273.1 6 - 7 793 104
XM_005260530.2 XP_005260587.1 6 - 7 761 104
XM_005260531.3 XP_005260588.1 6 - 7 761 104
XM_005260532.3 XP_005260589.1 7 - 8 983 104
XM_005260533.2 XP_005260590.1 6 - 7 787 104
XM_011529018.2 XP_011527320.1 7 - 8 1047 104
XM_011529020.2 XP_011527322.1 6 - 7 761 104
XM_017028029.1 XP_016883518.1 7 - 8 830 104
XM_017028030.1 XP_016883519.1 8 - 9 2188 104
XM_017028031.1 XP_016883520.1 8 - 9 2358 104
XM_017028032.1 XP_016883521.1 7 - 8 854 104
XM_017028033.1 XP_016883522.1 6 - 7 787 104
GenBank mRNA AB209315.1 - 6 - 7 770 104
AK225326.1 - - 752 104
AK309169.1 - 6 - 7 755 104
AK315238.1 - 6 - 7 731 104
BC015332.1 - 6 - 7 746 104
BC029511.1 - 6 - 7 766 104
BC058916.1 - 6 - 7 717 104
BC093768.1 - 6 - 7 735 104
CR933633.1 - 5 - 6 576 104
U18297.1 - 6 - 7 717 104
U60207.1 - 6 - 7 740 104

View Full Product Details