Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDX1
Entrez Gene ID
2230
Gene Name
ferredoxin 1
Gene Aliases
ADX, FDX, LOH11CR1D
Chromosome Location
Chr.11: 110429937 - 110464884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004109.4 NP_004100.1 1 - 2 369 166
GenBank mRNA AB096243.1 - 1 - 2 212 166
BC010284.1 - 1 - 2 338 166
BC017063.1 - 1 - 2 319 166
BT006681.1 - 1 - 2 185 166
J03548.1 - 1 - 2 317 166
M18003.1 - 1 - 2 212 166
M34788.1 - 1 - 2 203 166

View Full Product Details