Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDX1
Entrez Gene ID
2230
Gene Name
ferredoxin 1
Gene Aliases
ADX, FDX, LOH11CR1D
Chromosome Location
Chr.11: 110429937 - 110464884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004109.4 NP_004100.1 2 - 3 494 101
GenBank mRNA AB096243.1 - 2 - 3 337 101
BC010284.1 - 2 - 3 463 101
BC017063.1 - 2 - 3 444 101
BT006681.1 - 2 - 3 310 101
M18003.1 - 2 - 3 337 101
M34788.1 - 2 - 3 328 101

View Full Product Details