Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC5A
Entrez Gene ID
23601
Gene Name
C-type lectin domain family 5 member A
Gene Aliases
CLECSF5, MDL-1, MDL1
Chromosome Location
Chr.7: 141927356 - 141947009 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301167.1 NP_001288096.1 5 - 6 606 135
NM_013252.2 NP_037384.1 6 - 7 651 135
XM_011515995.2 XP_011514297.1 4 - 5 538 135
XM_017011915.1 XP_016867404.1 5 - 6 609 135
GenBank mRNA AF139768.1 - 6 - 7 602 135
AJ271684.1 - 6 - 7 651 135
AK292766.1 - 6 - 7 640 135
AK295200.1 - 5 - 6 543 135
AK307095.1 - 4 - 5 469 135
AK313703.1 - 6 - 7 640 135
BC093714.1 - 5 - 6 475 135
BC112099.1 - 5 - 6 475 135
BC113099.1 - 5 - 6 524 135

View Full Product Details