The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127182.1 | NP_001120654.1 | 2 - 3 | 370 | 123 |
NM_018131.4 | NP_060601.3 | 2 - 3 | 492 | 123 | |
GenBank mRNA | AB091343.1 | - | 2 - 3 | 481 | 123 |
AK001402.1 | - | 1 - 2 | 92 | 123 | |
AK223495.1 | - | 2 - 3 | 473 | 123 | |
AK315536.1 | - | 2 - 3 | 489 | 123 | |
AY788918.1 | - | 2 - 3 | 492 | 123 | |
BC008947.2 | - | 2 - 3 | 472 | 123 | |
BX640718.1 | - | 3 - 4 | 412 | 123 | |
DA545074.1 | - | 2 - 3 | 370 | 123 |