The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022168.3 | NP_071451.2 | 7 - 8 | 1931 | 114 |
GenBank mRNA | AF095844.1 | - | 7 - 8 | 1698 | 114 |
AK056293.1 | - | 1 - 2 | 108 | 114 | |
AK292941.1 | - | 8 - 9 | 1845 | 114 | |
AK314611.1 | - | 7 - 8 | 1926 | 114 | |
AY017378.1 | - | 7 - 8 | 1684 | 114 | |
BC078180.1 | - | 7 - 8 | 1753 | 114 | |
BC111750.1 | - | 7 - 8 | 1598 | 114 |