Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINT2
Entrez Gene ID
10653
Gene Name
serine peptidase inhibitor, Kunitz type 2
Gene Aliases
DIAR3, HAI-2, HAI2, Kop, PB
Chromosome Location
Chr.19: 38264458 - 38292614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166103.1 NP_001159575.1 5 - 6 853 64
NM_021102.3 NP_066925.1 6 - 7 1024 64
GenBank mRNA AB006534.1 - 6 - 7 901 64
AF027205.1 - 6 - 7 953 64
AK291532.1 - 6 - 7 839 64
AK297154.1 - 5 - 6 528 64
AX772830.1 - 5 - 6 735 64
BC001668.2 - 6 - 7 920 64
BC007705.2 - 6 - 7 64
BC011951.1 - 6 - 7 64
BC011955.1 - 6 - 7 64
BC012868.2 - 6 - 7 64
BT020115.1 - 6 - 7 589 64
BT020116.1 - 6 - 7 589 64
BT020117.1 - 6 - 7 589 64
CR407604.1 - 6 - 7 589 64
U78095.1 - 6 - 7 889 64

View Full Product Details