Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHODL
Entrez Gene ID
140578
Gene Name
chondrolectin
Gene Aliases
C21orf68, MT75, PRED12
Chromosome Location
Chr.21: 17819329 - 18267371 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204174.1 NP_001191103.1 5 - 6 745 116
NM_001204175.1 NP_001191104.1 5 - 6 711 116
NM_001204176.1 NP_001191105.1 6 - 7 811 116
NM_024944.2 NP_079220.2 5 - 6 1120 116
XM_011529457.2 XP_011527759.1 6 - 7 1117 116
GenBank mRNA AF257472.1 - 5 - 6 1120 116
AF523314.1 - 6 - 7 811 116
AK022689.1 - 6 - 7 993 116
AK296009.1 - 5 - 6 745 116
AK313725.1 - 5 - 6 768 116
AY358608.1 - 5 - 6 1106 116
BC009418.1 - 5 - 6 1090 116

View Full Product Details