The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032509.3 | NP_115898.2 | 5 - 6 | 434 | 97 |
GenBank mRNA | AF251062.1 | - | 5 - 6 | 409 | 97 |
AK027469.1 | - | 5 - 6 | 419 | 97 | |
AK225476.1 | - | 5 - 6 | 420 | 97 | |
AK314491.1 | - | 5 - 6 | 422 | 97 | |
BC028230.1 | - | 5 - 6 | 402 | 97 | |
BC039740.1 | - | 5 - 6 | 415 | 97 | |
BC050528.1 | - | 5 - 6 | 402 | 97 | |
BP281245.1 | - | 5 - 6 | 435 | 97 |