The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032509.3 | NP_115898.2 | 7 - 8 | 561 | 103 |
GenBank mRNA | AF251062.1 | - | 7 - 8 | 536 | 103 |
AK027469.1 | - | 7 - 8 | 546 | 103 | |
AK225476.1 | - | 7 - 8 | 547 | 103 | |
AK314491.1 | - | 7 - 8 | 103 | ||
BC028230.1 | - | 7 - 8 | 529 | 103 | |
BC039740.1 | - | 7 - 8 | 542 | 103 | |
BC050528.1 | - | 7 - 8 | 529 | 103 |