The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003149.2 | NP_003140.1 | 2 - 3 | 544 | 62 |
XM_011534036.2 | XP_011532338.1 | 3 - 4 | 577 | 62 | |
XM_011534037.2 | XP_011532339.1 | 2 - 3 | 543 | 62 | |
XM_011534038.2 | XP_011532340.1 | 3 - 4 | 582 | 62 | |
XM_017007083.1 | XP_016862572.1 | 2 - 3 | 416 | 62 | |
GenBank mRNA | AK302697.1 | - | 3 - 4 | 466 | 62 |
AK310425.1 | - | 3 - 4 | 582 | 62 | |
AK313493.1 | - | 2 - 3 | 539 | 62 | |
BC020221.1 | - | 2 - 3 | 536 | 62 | |
D86640.1 | - | 2 - 3 | 426 | 62 |