The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000441.1 | NP_000432.1 | 15 - 16 | 1937 | 143 |
XM_005250425.2 | XP_005250482.1 | 15 - 16 | 1920 | 143 | |
XM_017012318.1 | XP_016867807.1 | 13 - 14 | 1638 | 143 | |
GenBank mRNA | AF030880.1 | - | 15 - 16 | 1937 | 143 |
AK294388.1 | - | 6 - 7 | 816 | 143 | |
AK298543.1 | - | 5 - 6 | 560 | 143 | |
AK300604.1 | - | 6 - 7 | 632 | 143 |