The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016441.2 | NP_057525.1 | 7 - 8 | 1438 | 93 |
XM_005264357.4 | XP_005264414.1 | 6 - 7 | 1260 | 93 | |
XM_011532898.2 | XP_011531200.1 | 8 - 9 | 1559 | 93 | |
XM_011532899.2 | XP_011531201.1 | 8 - 9 | 1560 | 93 | |
XM_011532901.2 | XP_011531203.1 | 8 - 9 | 1560 | 93 | |
XM_017004258.1 | XP_016859747.1 | 7 - 8 | 1316 | 93 | |
XM_017004259.1 | XP_016859748.1 | 7 - 8 | 1577 | 93 | |
XM_017004260.1 | XP_016859749.1 | 6 - 7 | 1191 | 93 | |
XM_017004261.1 | XP_016859750.1 | 6 - 7 | 851 | 93 | |
XM_017004262.1 | XP_016859751.1 | 5 - 6 | 734 | 93 | |
GenBank mRNA | AB209139.1 | - | 7 - 8 | 1438 | 93 |
AF167706.1 | - | 7 - 8 | 1411 | 93 | |
AY358371.1 | - | 7 - 8 | 1739 | 93 | |
BC111989.1 | - | 7 - 8 | 1409 | 93 | |
BC113371.1 | - | 7 - 8 | 1409 | 93 | |
BX648755.1 | - | 1 - 2 | 87 | 93 |