Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGA1
Entrez Gene ID
1259
Gene Name
cyclic nucleotide gated channel alpha 1
Gene Aliases
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49
Chromosome Location
Chr.4: 47935015 - 48016718 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000087.3 NP_000078.2 3 - 4 266 129
XM_017007712.1 XP_016863201.1 3 - 4 321 129
GenBank mRNA AK292021.1 - 3 - 4 266 129
X64802.1 - 3 - 4 321 129

View Full Product Details