The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012471.2 | NP_036603.1 | 2 - 3 | 1290 | 77 |
XM_017029774.1 | XP_016885263.1 | 3 - 4 | 1580 | 77 | |
GenBank mRNA | AB209258.1 | - | 2 - 3 | 704 | 77 |
AF054568.1 | - | 2 - 3 | 1289 | 77 | |
BC137271.1 | - | 2 - 3 | 1130 | 77 | |
BC137274.1 | - | 2 - 3 | 1130 | 77 |