The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012471.2 | NP_036603.1 | 5 - 6 | 2291 | 69 |
XM_017029774.1 | XP_016885263.1 | 6 - 7 | 2581 | 69 | |
GenBank mRNA | AB209258.1 | - | 5 - 6 | 1705 | 69 |
AF054568.1 | - | 5 - 6 | 2290 | 69 | |
BC137271.1 | - | 5 - 6 | 2131 | 69 | |
BC137274.1 | - | 5 - 6 | 2131 | 69 |