The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012471.2 | NP_036603.1 | 7 - 8 | 2816 | 81 |
XM_017029774.1 | XP_016885263.1 | 8 - 9 | 3106 | 81 | |
GenBank mRNA | AB209258.1 | - | 7 - 8 | 2230 | 81 |
AF054568.1 | - | 7 - 8 | 2815 | 81 | |
BC137271.1 | - | 7 - 8 | 2656 | 81 | |
BC137274.1 | - | 7 - 8 | 2656 | 81 |