The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171713.1 | NP_001165184.1 | 11 - 12 | 755 | 74 |
NM_012157.3 | NP_036289.3 | 12 - 13 | 962 | 74 | |
XM_005265014.2 | XP_005265071.1 | 12 - 13 | 982 | 74 | |
XM_005265015.3 | XP_005265072.1 | 12 - 13 | 1202 | 74 | |
XM_005265016.2 | XP_005265073.1 | 12 - 13 | 946 | 74 | |
XM_005265017.3 | XP_005265074.1 | 12 - 13 | 978 | 74 | |
XM_011533559.2 | XP_011531861.1 | 13 - 14 | 1147 | 74 | |
XM_017006092.1 | XP_016861581.1 | 13 - 14 | 1039 | 74 | |
XM_017006093.1 | XP_016861582.1 | 12 - 13 | 1029 | 74 | |
XM_017006094.1 | XP_016861583.1 | 13 - 14 | 1062 | 74 | |
GenBank mRNA | AF176518.1 | - | 12 - 13 | 897 | 74 |
AF186273.1 | - | 12 - 13 | 912 | 74 | |
AK001271.1 | - | 12 - 13 | 962 | 74 | |
AK001438.1 | - | 12 - 13 | 930 | 74 | |
AK294209.1 | - | 9 - 10 | 713 | 74 | |
AK298967.1 | - | 11 - 12 | 724 | 74 | |
AK299665.1 | - | 13 - 14 | 1045 | 74 | |
AK302218.1 | - | 14 - 15 | 1190 | 74 | |
AK303764.1 | - | 10 - 11 | 822 | 74 | |
AK316229.1 | - | 12 - 13 | 983 | 74 | |
AK316383.1 | - | 13 - 14 | 1102 | 74 | |
AL049953.1 | - | 5 - 6 | 372 | 74 | |
BC031556.1 | - | 12 - 13 | 906 | 74 | |
CR457121.1 | - | 12 - 13 | 891 | 74 |