The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171713.1 | NP_001165184.1 | 8 - 9 | 519 | 80 |
NM_012157.3 | NP_036289.3 | 9 - 10 | 726 | 80 | |
XM_005265014.2 | XP_005265071.1 | 9 - 10 | 746 | 80 | |
XM_005265015.3 | XP_005265072.1 | 9 - 10 | 966 | 80 | |
XM_005265016.2 | XP_005265073.1 | 9 - 10 | 710 | 80 | |
XM_005265017.3 | XP_005265074.1 | 9 - 10 | 742 | 80 | |
XM_011533559.2 | XP_011531861.1 | 10 - 11 | 911 | 80 | |
XM_017006092.1 | XP_016861581.1 | 10 - 11 | 803 | 80 | |
XM_017006093.1 | XP_016861582.1 | 9 - 10 | 793 | 80 | |
XM_017006094.1 | XP_016861583.1 | 10 - 11 | 826 | 80 | |
GenBank mRNA | AF174589.1 | - | 10 - 11 | 697 | 80 |
AF176518.1 | - | 9 - 10 | 661 | 80 | |
AF186273.1 | - | 9 - 10 | 676 | 80 | |
AK001271.1 | - | 9 - 10 | 726 | 80 | |
AK001438.1 | - | 9 - 10 | 694 | 80 | |
AK294209.1 | - | 6 - 7 | 477 | 80 | |
AK297534.1 | - | 9 - 10 | 710 | 80 | |
AK298967.1 | - | 8 - 9 | 488 | 80 | |
AK299665.1 | - | 10 - 11 | 809 | 80 | |
AK302218.1 | - | 11 - 12 | 954 | 80 | |
AK303764.1 | - | 7 - 8 | 586 | 80 | |
AK316229.1 | - | 9 - 10 | 747 | 80 | |
AK316383.1 | - | 10 - 11 | 866 | 80 | |
AL049953.1 | - | 2 - 3 | 136 | 80 | |
BC031556.1 | - | 9 - 10 | 670 | 80 | |
CR457121.1 | - | 9 - 10 | 655 | 80 |