Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH6
Entrez Gene ID
347365
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 6
Gene Aliases
ITIH5L, UNQ6369, dJ14O9.1
Chromosome Location
Chr.X: 54748899 - 54798240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198510.2 NP_940912.1 6 - 7 925 73
GenBank mRNA AY358170.1 - 6 - 7 925 73

View Full Product Details