Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC10
Entrez Gene ID
23008
Gene Name
kelch domain containing 10
Gene Aliases
slim
Chromosome Location
Chr.7: 130070509 - 130135720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014997.3 NP_055812.1 7 - 8 1071 122
XM_005250233.4 XP_005250290.1 6 - 7 960 122
GenBank mRNA BC009977.2 - 5 - 6 484 122
BC036464.2 - 7 - 8 1053 122
BC044884.1 - 7 - 8 1073 122
D87454.1 - 7 - 8 813 122

View Full Product Details