The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033054.2 | NP_149043.2 | 10 - 11 | 1481 | 68 |
XM_017012503.1 | XP_016867992.1 | 9 - 10 | 1240 | 68 | |
XM_017012504.1 | XP_016867993.1 | 10 - 11 | 1476 | 68 | |
GenBank mRNA | AB290179.1 | - | 10 - 11 | 1441 | 68 |
AF380932.1 | - | 9 - 10 | 1241 | 68 | |
AF380933.1 | - | 9 - 10 | 1241 | 68 | |
AK074050.1 | - | 9 - 10 | 4324 | 68 | |
AK074135.1 | - | 6 - 7 | 761 | 68 | |
AK097912.1 | - | 10 - 11 | 1472 | 68 | |
AK131089.1 | - | 5 - 6 | 3598 | 68 | |
AL832302.1 | - | 9 - 10 | 1236 | 68 | |
BC113544.1 | - | 10 - 11 | 1428 | 68 | |
BC117241.1 | - | 10 - 11 | 1428 | 68 | |
BI910371.1 | - | 2 - 3 | 179 | 68 |