The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015458.3 | NP_056273.2 | 1 - 2 | 579 | 66 |
XM_011543830.2 | XP_011542132.1 | 1 - 2 | 243 | 66 | |
XM_017013753.1 | XP_016869242.1 | 1 - 2 | 537 | 66 | |
GenBank mRNA | AJ297823.1 | - | 1 - 2 | 263 | 66 |
AK000444.1 | - | 1 - 2 | 263 | 66 | |
AK223525.1 | - | 1 - 2 | 263 | 66 | |
AK303543.1 | - | 1 - 2 | 252 | 66 | |
BC022003.1 | - | 1 - 2 | 222 | 66 | |
BC034990.2 | - | 1 - 2 | 580 | 66 | |
BC093788.1 | - | 1 - 2 | 224 | 66 | |
BC101623.1 | - | 1 - 2 | 224 | 66 | |
BX538176.1 | - | 1 - 2 | 245 | 66 |