Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR9
Entrez Gene ID
66036
Gene Name
myotubularin related protein 9
Gene Aliases
C8orf9, LIP-STYX, MTMR8
Chromosome Location
Chr.8: 11284491 - 11328146 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015458.3 NP_056273.2 4 - 5 985 89
XM_011543830.2 XP_011542132.1 5 - 6 701 89
XM_011543831.2 XP_011542133.1 2 - 3 257 89
XM_017013753.1 XP_016869242.1 4 - 5 943 89
GenBank mRNA AJ297823.1 - 4 - 5 669 89
AK294463.1 - 4 - 5 576 89
AK303543.1 - 4 - 5 601 89
BC022003.1 - 4 - 5 628 89
BC034990.2 - 4 - 5 986 89
BC093788.1 - 4 - 5 630 89
BC101623.1 - 4 - 5 630 89
BX538176.1 - 5 - 6 703 89

View Full Product Details