The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015458.3 | NP_056273.2 | 8 - 9 | 1725 | 72 |
XM_011543830.2 | XP_011542132.1 | 9 - 10 | 1441 | 72 | |
XM_011543831.2 | XP_011542133.1 | 6 - 7 | 997 | 72 | |
GenBank mRNA | AJ297823.1 | - | 8 - 9 | 1409 | 72 |
AK000444.1 | - | 8 - 9 | 1409 | 72 | |
AK223525.1 | - | 8 - 9 | 1409 | 72 | |
AK294463.1 | - | 8 - 9 | 1316 | 72 | |
AK303543.1 | - | 8 - 9 | 1341 | 72 | |
BC022003.1 | - | 8 - 9 | 1368 | 72 | |
BC034990.2 | - | 8 - 9 | 1726 | 72 | |
BC093788.1 | - | 8 - 9 | 1370 | 72 | |
BC101623.1 | - | 8 - 9 | 1370 | 72 | |
BX538176.1 | - | 9 - 10 | 1443 | 72 |