The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193308.1 | NP_001180237.1 | 12 - 13 | 1445 | 94 |
NM_032872.2 | NP_116261.1 | 12 - 13 | 1409 | 94 | |
XM_005246022.3 | XP_005246079.1 | 12 - 13 | 1600 | 94 | |
XM_006710990.1 | XP_006711053.1 | 12 - 13 | 1564 | 94 | |
XM_006710991.1 | XP_006711054.1 | 11 - 12 | 1199 | 94 | |
GenBank mRNA | AK027902.1 | - | 12 - 13 | 1379 | 94 |
AK096437.1 | - | 10 - 11 | 2006 | 94 | |
AK097172.1 | - | 5 - 6 | 2162 | 94 | |
AK293038.1 | - | 11 - 12 | 1711 | 94 | |
AY037157.1 | - | 12 - 13 | 1372 | 94 | |
BC009224.2 | - | 10 - 11 | 976 | 94 | |
BC015764.1 | - | 11 - 12 | 1188 | 94 | |
BC035725.1 | - | 12 - 13 | 1352 | 94 |