The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320821.1 | NP_001307750.1 | 22 - 23 | 2537 | 83 |
NM_001320822.1 | NP_001307751.1 | 20 - 21 | 2059 | 83 | |
NM_007368.3 | NP_031394.2 | 20 - 21 | 2071 | 83 | |
XM_011534841.2 | XP_011533143.1 | 20 - 21 | 2136 | 83 | |
XM_011534842.2 | XP_011533144.1 | 20 - 21 | 2196 | 83 | |
XM_017020438.1 | XP_016875927.1 | 22 - 23 | 2485 | 83 | |
XM_017020439.1 | XP_016875928.1 | 21 - 22 | 2282 | 83 | |
GenBank mRNA | AK128094.1 | - | 22 - 23 | 2506 | 83 |
AK289746.1 | - | 20 - 21 | 1977 | 83 | |
AK294913.1 | - | 20 - 21 | 2044 | 83 | |
AK313795.1 | - | 20 - 21 | 2005 | 83 | |
BC038456.1 | - | 20 - 21 | 2068 | 83 | |
BC047242.1 | - | 20 - 21 | 2059 | 83 | |
X89399.1 | - | 20 - 21 | 1983 | 83 |