The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320821.1 | NP_001307750.1 | 24 - 25 | 2850 | 62 |
NM_001320822.1 | NP_001307751.1 | 22 - 23 | 2372 | 62 | |
NM_007368.3 | NP_031394.2 | 22 - 23 | 2384 | 62 | |
XM_011534841.2 | XP_011533143.1 | 22 - 23 | 2449 | 62 | |
XM_011534842.2 | XP_011533144.1 | 22 - 23 | 2509 | 62 | |
XM_017020438.1 | XP_016875927.1 | 24 - 25 | 2798 | 62 | |
XM_017020439.1 | XP_016875928.1 | 23 - 24 | 2595 | 62 | |
GenBank mRNA | AK128094.1 | - | 24 - 25 | 2819 | 62 |
AK289746.1 | - | 22 - 23 | 2290 | 62 | |
AK294913.1 | - | 22 - 23 | 2357 | 62 | |
AK313795.1 | - | 22 - 23 | 2318 | 62 | |
BC038456.1 | - | 22 - 23 | 2381 | 62 | |
BC047242.1 | - | 22 - 23 | 2372 | 62 | |
X89399.1 | - | 22 - 23 | 2296 | 62 |