The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025580.1 | NP_001020751.1 | 4 - 5 | 853 | 135 |
NM_001171689.1 | NP_001165160.1 | 7 - 8 | 1151 | 135 | |
NM_015365.2 | NP_056180.1 | 5 - 6 | 964 | 135 | |
GenBank mRNA | AJ007014.1 | - | 5 - 6 | 1051 | 135 |
AK001703.1 | - | 5 - 6 | 487 | 135 | |
AK091430.1 | - | 7 - 8 | 1151 | 135 | |
AK314596.1 | - | 5 - 6 | 895 | 135 | |
BC060813.1 | - | 4 - 5 | 786 | 135 | |
BX648797.1 | - | 4 - 5 | 815 | 135 | |
KU178496.1 | - | 4 - 5 | 772 | 135 | |
KU178497.1 | - | 5 - 6 | 883 | 135 |