The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000384.2 | NP_000375.2 | 24 - 25 | 3975 | 100 |
GenBank mRNA | HM487065.1 | - | 24 - 25 | 3847 | 100 |
J02610.1 | - | 24 - 25 | 3925 | 100 | |
M14081.1 | - | 24 - 25 | 3847 | 100 | |
X04506.1 | - | 24 - 25 | 3975 | 100 | |
X04714.1 | - | 24 - 25 | 3966 | 100 |