The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012135.2 | NP_036267.1 | 1 - 2 | 124 | 71 |
GenBank mRNA | BC001261.1 | - | 1 - 2 | 102 | 71 |
BI488606.1 | - | - | 125 | 71 | |
Y18503.1 | - | 1 - 2 | 92 | 71 |