The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008396.2 | NP_001008397.1 | 4 - 5 | 445 | 78 |
NM_007086.3 | NP_009017.1 | 5 - 6 | 538 | 78 | |
XM_006720012.2 | XP_006720075.1 | 5 - 6 | 536 | 78 | |
XM_011536373.2 | XP_011534675.1 | 4 - 5 | 449 | 78 | |
XM_017020940.1 | XP_016876429.1 | 4 - 5 | 449 | 78 | |
XM_017020941.1 | XP_016876430.1 | 2 - 3 | 259 | 78 | |
GenBank mRNA | AJ006266.1 | - | 4 - 5 | 487 | 78 |
AK293005.1 | - | 5 - 6 | 505 | 78 | |
BC000622.2 | - | 5 - 6 | 503 | 78 | |
BC005040.1 | - | 5 - 6 | 503 | 78 | |
BC043349.1 | - | 5 - 6 | 526 | 78 | |
BC063041.1 | - | 5 - 6 | 514 | 78 | |
DA189308.1 | - | 4 - 5 | 445 | 78 |