Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDHD1
Entrez Gene ID
11169
Gene Name
WD repeat and HMG-box DNA binding protein 1
Gene Aliases
AND-1, AND1, CHTF4, CTF4
Chromosome Location
Chr.14: 54938938 - 55027105 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008396.2 NP_001008397.1 6 - 7 602 154
NM_007086.3 NP_009017.1 7 - 8 695 154
XM_006720012.2 XP_006720075.1 7 - 8 693 154
XM_011536373.2 XP_011534675.1 6 - 7 606 154
XM_017020940.1 XP_016876429.1 6 - 7 606 154
XM_017020941.1 XP_016876430.1 4 - 5 416 154
GenBank mRNA AJ006266.1 - 6 - 7 644 154
AK293005.1 - 7 - 8 662 154
BC000622.2 - 7 - 8 660 154
BC005040.1 - 7 - 8 660 154
BC043349.1 - 7 - 8 683 154
BC063041.1 - 7 - 8 671 154

View Full Product Details