Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HIF3A
Entrez Gene ID
64344
Gene Name
hypoxia inducible factor 3 alpha subunit
Gene Aliases
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
Chromosome Location
Chr.19: 46297046 - 46343433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022462.4 NP_071907.4 10 - 11 1401 65
NM_152794.3 NP_690007.1 11 - 12 1497 65
NM_152795.3 NP_690008.2 11 - 12 1465 65
XM_005259152.4 XP_005259209.1 12 - 13 1593 65
XM_005259153.3 XP_005259210.2 9 - 10 1264 65
XM_017027136.1 XP_016882625.1 12 - 13 1593 65
XM_017027138.1 XP_016882627.1 11 - 12 1463 65
GenBank mRNA AB054067.1 - 11 - 12 1428 65
AB295039.1 - 11 - 12 1446 65
AF463492.1 - 11 - 12 1460 65
AK021653.1 - 12 - 13 1491 65
AK027725.1 - 11 - 12 1446 65
AK297828.1 - 10 - 11 1387 65
AK299699.1 - 10 - 11 1311 65
BC080551.1 - 12 - 13 1436 65

View Full Product Details