The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317960.1 | NP_001304889.1 | 11 - 12 | 1340 | 154 |
NM_012255.4 | NP_036387.2 | 11 - 12 | 1180 | 154 | |
XM_017027722.1 | XP_016883211.1 | 11 - 12 | 1335 | 154 | |
XM_017027723.1 | XP_016883212.1 | 11 - 12 | 1109 | 154 | |
XM_017027724.1 | XP_016883213.1 | 11 - 12 | 1335 | 154 | |
GenBank mRNA | AF064257.1 | - | 11 - 12 | 1133 | 154 |
AF152169.1 | - | 11 - 12 | 1083 | 154 | |
AK001573.1 | - | 11 - 12 | 1150 | 154 | |
AK302846.1 | - | 11 - 12 | 1044 | 154 | |
AK303312.1 | - | 10 - 11 | 991 | 154 | |
AK314602.1 | - | 11 - 12 | 1119 | 154 | |
AL136841.1 | - | 11 - 12 | 1106 | 154 | |
AY382900.1 | - | 10 - 11 | 995 | 154 |