The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317960.1 | NP_001304889.1 | 6 - 7 | 852 | 141 |
NM_012255.4 | NP_036387.2 | 6 - 7 | 692 | 141 | |
XM_017027722.1 | XP_016883211.1 | 6 - 7 | 847 | 141 | |
XM_017027723.1 | XP_016883212.1 | 6 - 7 | 621 | 141 | |
XM_017027724.1 | XP_016883213.1 | 6 - 7 | 847 | 141 | |
GenBank mRNA | AF064257.1 | - | 6 - 7 | 645 | 141 |
AF152169.1 | - | 6 - 7 | 595 | 141 | |
AK001573.1 | - | 6 - 7 | 662 | 141 | |
AK302846.1 | - | 6 - 7 | 556 | 141 | |
AK303312.1 | - | 5 - 6 | 503 | 141 | |
AK314602.1 | - | 6 - 7 | 631 | 141 | |
AL136841.1 | - | 6 - 7 | 618 | 141 | |
AY382900.1 | - | 5 - 6 | 507 | 141 |