The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317960.1 | NP_001304889.1 | 7 - 8 | 928 | 114 |
NM_012255.4 | NP_036387.2 | 7 - 8 | 768 | 114 | |
XM_017027722.1 | XP_016883211.1 | 7 - 8 | 923 | 114 | |
XM_017027723.1 | XP_016883212.1 | 7 - 8 | 697 | 114 | |
XM_017027724.1 | XP_016883213.1 | 7 - 8 | 923 | 114 | |
GenBank mRNA | AF152169.1 | - | 7 - 8 | 671 | 114 |
AK001573.1 | - | 7 - 8 | 738 | 114 | |
AK302846.1 | - | 7 - 8 | 632 | 114 | |
AK303312.1 | - | 6 - 7 | 579 | 114 | |
AK314602.1 | - | 7 - 8 | 707 | 114 | |
AL136841.1 | - | 7 - 8 | 694 | 114 | |
AY382900.1 | - | 6 - 7 | 583 | 114 |