Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANGEL1
Entrez Gene ID
23357
Gene Name
angel homolog 1
Gene Aliases
Ccr4e, KIAA0759
Chromosome Location
Chr.14: 76786473 - 76812947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015305.3 NP_056120.2 4 - 5 1057 63
XM_017021115.1 XP_016876604.1 4 - 5 1064 63
GenBank mRNA AB018302.1 - 4 - 5 954 63
AL137268.1 - 4 - 5 955 63
BC028714.2 - 4 - 5 1004 63

View Full Product Details