The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005113.3 | NP_005104.3 | 7 - 8 | 1746 | 71 |
XM_011537420.2 | XP_011535722.1 | 7 - 8 | 1746 | 71 | |
GenBank mRNA | AF085199.1 | - | 7 - 8 | 1621 | 71 |
AJ132949.1 | - | 6 - 7 | 1490 | 71 | |
AK293077.1 | - | 7 - 8 | 1649 | 71 | |
AK303789.1 | - | 8 - 9 | 1379 | 71 | |
AY644768.1 | - | 6 - 7 | 1526 | 71 | |
BC023021.1 | - | 7 - 8 | 1654 | 71 |