The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005113.3 | NP_005104.3 | 9 - 10 | 1974 | 109 |
XM_011537420.2 | XP_011535722.1 | 9 - 10 | 1974 | 109 | |
GenBank mRNA | AF085199.1 | - | 9 - 10 | 1849 | 109 |
AK293077.1 | - | 9 - 10 | 1877 | 109 | |
AK303789.1 | - | 10 - 11 | 1607 | 109 | |
AY644768.1 | - | 8 - 9 | 1754 | 109 | |
BC023021.1 | - | 9 - 10 | 1882 | 109 |