The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244984.1 | NP_001231913.1 | 6 - 7 | 894 | 80 |
NM_005065.5 | NP_005056.3 | 6 - 7 | 894 | 80 | |
XM_005267988.2 | XP_005268045.1 | 6 - 7 | 777 | 80 | |
XM_005267989.3 | XP_005268046.1 | 6 - 7 | 897 | 80 | |
GenBank mRNA | AB020335.1 | - | 6 - 7 | 822 | 80 |
AF052059.1 | - | 6 - 7 | 870 | 80 | |
AK075511.1 | - | 6 - 7 | 873 | 80 | |
AY358651.1 | - | 6 - 7 | 866 | 80 |