The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321643.1 | NP_001308572.1 | 20 - 21 | 2545 | 151 |
NM_004850.4 | NP_004841.2 | 20 - 21 | 3001 | 151 | |
XM_005246190.3 | XP_005246247.1 | 20 - 21 | 3035 | 151 | |
XM_011510417.2 | XP_011508719.1 | 20 - 21 | 2500 | 151 | |
XM_017005378.1 | XP_016860867.1 | 20 - 21 | 3032 | 151 | |
XM_017005379.1 | XP_016860868.1 | 20 - 21 | 2430 | 151 | |
GenBank mRNA | AB014519.1 | - | 20 - 21 | 3006 | 151 |
AK299296.1 | - | 6 - 7 | 1008 | 151 | |
AK310358.1 | - | 16 - 17 | 2296 | 151 | |
BC111801.1 | - | 7 - 8 | 1108 | 151 | |
D87931.1 | - | 20 - 21 | 2552 | 151 |