The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000329.2 | NP_000320.1 | 4 - 5 | 398 | 110 |
XM_017002027.1 | XP_016857516.1 | 3 - 4 | 610 | 110 | |
GenBank mRNA | AK289925.1 | - | 4 - 5 | 389 | 110 |
BC075035.2 | - | 4 - 5 | 390 | 110 | |
BC075036.2 | - | 4 - 5 | 393 | 110 | |
U18991.1 | - | 4 - 5 | 398 | 110 |