Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
Chromosome Location
Chr.6: 49430360 - 49463328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000255.3 NP_000246.2 3 - 4 1016 111
XM_005249143.3 XP_005249200.1 3 - 4 921 111
GenBank mRNA AK292568.1 - 3 - 4 992 111
AK312611.1 - 3 - 4 991 111
BC016282.1 - 3 - 4 879 111
BT007434.1 - 2 - 3 751 111
BX647789.1 - 3 - 4 841 111
KF030882.1 - 2 - 3 751 111
KU178201.1 - 2 - 3 751 111
KU178202.1 - 2 - 3 751 111
M65131.1 - 3 - 4 827 111

View Full Product Details