Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
Chromosome Location
Chr.6: 49430360 - 49463328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000255.3 NP_000246.2 4 - 5 1179 95
XM_005249143.3 XP_005249200.1 4 - 5 1084 95
GenBank mRNA AK292568.1 - 4 - 5 1155 95
AK312611.1 - 4 - 5 1154 95
BC016282.1 - 4 - 5 1042 95
BT007434.1 - 3 - 4 914 95
BX647789.1 - 4 - 5 1004 95
KF030882.1 - 3 - 4 914 95
KU178201.1 - 3 - 4 914 95
KU178202.1 - 3 - 4 914 95
M65131.1 - 4 - 5 990 95

View Full Product Details